EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
Center punch and whole spot bioanalysis of apixaban in human dried blood spot samples by UHPLC-MS/MS.
PMID 25746754