EMAIL THIS PAGE TO A FRIEND
PLoS Pathogens
The role of Misshapen NCK-related kinase (MINK), a novel Ste20 family kinase, in the IRES-mediated protein translation of human enterovirus 71.
PMID 25747578