EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Heart and circulatory physiology
Mineralocorticoid receptor blockade prevents Western diet-induced diastolic dysfunction in female mice.
PMID 25747754
Abstract
Overnutrition/obesity predisposes individuals, particularly women, to diastolic dysfunction (DD), an independent predictor of future cardiovascular disease. We examined whether low-dose spironolactone (Sp) prevents DD associated with consumption of a Western Diet (WD) high in fat, fructose, and sucrose. Female C57BL6J mice were fed a WD with or without Sp (1 mg·kg(-1)·day(-1)). After 4 mo on the WD, mice exhibited increased body weight and visceral fat, but similar blood pressures, compared with control diet-fed mice. Sp prevented the development of WD-induced DD, as indicated by decreased isovolumic relaxation time and an improvement in myocardial performance (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7389
3-Nitro-L-tyrosine, crystalline
C9H10N2O5