EMAIL THIS PAGE TO A FRIEND
Biological & pharmaceutical bulletin
Biotransformation of (-)-epigallocatechin and (-)-gallocatechin by intestinal bacteria involved in isoflavone metabolism.
PMID 25747993
Abstract
Four isoflavone-metabolizing bacteria were tested for their abilities to degrade (-)-epigallocatechin (EGC) and its isomer (-)-gallocatechin (GC). Biotransformation of both EGC and GC was observed with Adlercreutzia equolifaciens JCM 14793, Asaccharobacter celatus JCM 14811, and Slackia equolifaciens JCM 16059, but not Slackia isoflavoniconvertens JCM 16137. With respect to the degradation of EGC, strain JCM 14793 only catalyzed 4'-dehydroxylation to produce 4'-dehydroxylated EGC (7). Strain JCM 14811 mainly produced 7, along with a slight formation of the C ring-cleaving product 1-(3,4,5-trihydroxyphenyl)-3-(2,4,6-trihydroxyphenyl)propan-2-ol (1). Strain JCM 16059 catalyzed only C ring cleavage to form 1. Interestingly, the presence of hydrogen promoted the bioconversion of EGC by these bacteria. In addition, strain JCM 14811 revealed the ability to catalyze 4'-dehydroxylation of 1 to yield 1-(3,5-dihydroxyphenyl)-3-(2,4,6-trihydroxyphenyl)propan-2-ol (2) in the presence of hydrogen. In the case of GC, strain JCM 14793 mainly produced C ring-cleaving product (1) with only a very small amount of 4'-dehydroxylated GC (8), while Strain JCM 14811 only catalyzed 4'-dehydroxylation to form 8. Strain JCM 16059 formed 1. The bioconversion of GC by the three strains was stimulated by hydrogen. Strain JCM 14793 showed the ability to convert 1 into 2 in the presence of hydrogen as did strain JCM 14811. Furthermore, strains JCM 14793 and JCM 14811 were found to have the ability to catalyze p-dehydroxylation of the pyrogallol moiety in the EGC metabolites 4-hydroxy-5-(3,4,5-trihydroxyphenyl)valeric acid (3) and 5-(3,4,5-trihydroxyphenyl)-γ-valerolactone (4), and this ability was enhanced by the presence of hydrogen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31589
Diethanolamine, BioUltra, ≥99.5% (GC)
C4H11NO2
16957
Diethanolamine, analytical standard
C4H11NO2
31590
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
C4H11NO2
398179
Diethanolamine, ACS reagent, ≥98.5%
C4H11NO2
PHR1220
Diethanolamine, Pharmaceutical Secondary Standard; Certified Reference Material
C4H11NO2
721948 Methanol-d4, NMR reference standard, 99.8 atom % D, NMR tube size 5 mm × 8 in.
CD4O
721921 Methanol-d4, NMR reference standard, 99.8 atom % D, NMR tube size 3 mm × 8 in.
CD4O
444758
Methanol-d4, "100%", 99.96 atom % D
CD4O
535435
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
CD4O
151947
Methanol-d4, ≥99.8 atom % D
CD4O
441384
Methanol-d4, ≥99.8 atom % D
CD4O
343803
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
CD4O
530530
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
CD4O
611646
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
CD4O
439029
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
CD4O
417653
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
CD4O
422878
Methanol-d4, 99 atom % D
CD4O
569534
Methanol-d4, anhydrous, ≥99.8 atom % D
CD4O
570729
Methanol-d4, ≥99.8 atom % D, anhydrous
CD4O
Y0001183
Trolamine impurity B, European Pharmacopoeia (EP) Reference Standard
C4H11NO2