EMAIL THIS PAGE TO A FRIEND
Dental materials journal
Ag-loaded mesoporous bioactive glasses against Enterococcus faecalis biofilm in root canal of human teeth.
PMID 25748459
Abstract
Ag-loaded mesoporous bioactive glass (Ag-MBG) powders were synthesized and characterized. The ions release of Ag-MBGs in Tris-HCl and the pH stability of simulated body fluids after immersing Ag-MBGs were tested. Root canals were inoculated with Enterococcus faecalis for 4 weeks, and the antibacterial activity of MBGs, Ag-MBGs and calcium hydroxide against E. faecalis biofilm were evaluated. Results showed that Ag-MBGs possessed highly ordered mesoporous structure with silver nanoparticles deposited in the mesopores, which enabled a sustained Ag ions released. The biofilms treated with Ag-MBGs showed a significant structural disruption compared with MBGs. These results indicated that Ag-MBGs possess a potent antibacterial effect against E.faecalis biofilm in root canal, and the antibacterial activity was induced by the release of Ag ions from Ag-MBGs.