EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
High-performance liquid chromatographic enantioseparation of unusual amino acid derivatives with axial chirality on polysaccharide-based chiral stationary phases.
PMID 25748541