EMAIL THIS PAGE TO A FRIEND
Chemical & Pharmaceutical Bulletin
Preparation of methacrylic acid copolymer S nano-fibers using a solvent-based electrospinning method and their application in pharmaceutical formulations.
PMID 25748778