EMAIL THIS PAGE TO A FRIEND
Immunopharmacology and immunotoxicology
Attenuating effects of coenzyme Q10 and amlodipine in ulcerative colitis model in rats.
PMID 25753843
Abstract
Ulcerative colitis is a chronic inflammatory bowel disease. Recent studies reported a pivotal role of elevated intracellular calcium in this disorder. Coenzyme Q10 (CoQ10) and amlodipine are known to maintain cellular energy, decrease intracellular calcium concentration in addition to their antioxidant and anti-inflammatory properties. The aim of this study was to evaluate the possible protective effects of CoQ10, amlodipine and their combination on ulcerative colitis. Colitis was induced in rats by intracolonic injection of 3% acetic acid. CoQ10 (10 mg/kg), amlodipine (3 mg/kg) and their combination were administered for 8 consecutive days before induction of colitis. Our results showed that administration of CoQ10, amlodipine and their combination decreased colon tissue malondialdehyde (MDA), tumor necrosis factor-α (TNF-α), interleukin-1β (IL-1β), prostaglandin E2 (PGE2), myeloperoxidase (MPO) and heat shock protein (HSP70) levels induced by intracolonic injection of acetic acid and restored many of the colon structure in histological examination. On the other hand, they increased superoxide dismutase (SOD) activity, adenosine-5'-triphosphate (ATP) and interleukin-10 (IL-10) colonic contents. Administration of either CoQ10 or amlodipine was found to protect against acetic acid-induced colitis. Moreover, their combination was more effective than individual administration of either of them. The protective effect of CoQ10 and amlodipine may be in part via their antioxidant, anti-inflammatory and energy restoration properties.