EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
Inhibition of protein synthesis leading to unfolded protein response is the major event in abrin-mediated apoptosis.
PMID 25753921
Abstract
Abrin obtained from the plant Abrus precatorius inhibits protein synthesis and also triggers apoptosis in cells. Previous studies from our laboratory suggested a link between these two events. Using an active site mutant of abrin A-chain which exhibits 225-fold lower protein synthesis inhibitory activity than the wild-type abrin A-chain, we demonstrate in this study that inhibition of protein synthesis induced by abrin is the major factor triggering unfolded protein response leading to apoptosis. Since abrin A-chain requires the B-chain for internalization into cells, the wild-type and mutant recombinant abrin A-chains were conjugated to native ricin B-chain to generate hybrid toxins, and the toxic effects of the two conjugates were compared. The rate of inhibition of protein synthesis mediated by the mutant ricin B-rABRA (R167L) conjugate was slower than that of the wild-type ricin B-rABRA conjugate as expected. The mutant conjugate activated p38MAPK and caspase-3 similar to its wild-type counterpart although at later time points. Overall, these results confirm that inhibition of protein synthesis is the major event contributing to abrin-mediated apoptosis.