EMAIL THIS PAGE TO A FRIEND
Journal of cellular and molecular medicine
Association of vitamin D3 with alveolar bone regeneration in dogs.
PMID 25753943
Abstract
Designed sockets prepared on the mandibles of nine Beagle dogs were divided into three groups: Calcitriol +Alloplast, Alloplast and Empty. Five of the nine dogs received Vit.D3 and calcium supplement (Vit.D/Ca group), while the other four dogs without supplements were assigned to Non-Vit.D/Ca group. After 4xa0weeks, the extent of vertical ridge resorption (VRR), bone density (density), new bone formation (NBF) and implant stability quotient (ISQ) were measured. Following systemic Vit.D/Ca administration, the Empty subgroup showed significant differences from the Calcitriol + Alloplast subgroup on variants NBF/Density/VRR and the Alloplast subgroup on items NBF/Density/ISQ/VRR. Alternatively, the Calcitriol + Alloplast subgroup revealed higher values of NBF/Density/ISQ (Pxa0