EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
Development and validation of an ultra-high performance liquid chromatography-tandem mass spectrometry method to measure creatinine in human urine.
PMID 25756209