EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Structural, thermal, and anti-inflammatory properties of a novel pectic polysaccharide from alfalfa (Medicago sativa L.) stem.
PMID 25756601