EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Schinus terebinthifolius scale-up countercurrent chromatography (Part I): High performance countercurrent chromatography fractionation of triterpene acids with off-line detection using atmospheric pressure chemical ionization mass spectrometry.
PMID 25757818