EMAIL THIS PAGE TO A FRIEND
Nature Communications
Artificial human Met agonists based on macrocycle scaffolds.
PMID 25758345
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SRP3027 EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
11376454001 Epidermal Growth Factor, human (hEGF), recombinant (E. coli)
E5036
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
SRP4037 FGF-2 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
SRP2092
FGF-2 human, recombinant, expressed in insect cells, ≥85% (SDS-PAGE)
SRP3052
HB-EGF human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), cell culture tested
E9644
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
SRP3056 HVEM-Fc human, recombinant, expressed in Hi-5 Insect cells, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
1316004
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
C21H30O5
PHR1014
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
C21H30O5
31719
Hydrocortisone, VETRANAL, analytical standard
C21H30O5
H1300000
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
C21H30O5
BP576
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
C21H30O5
Y0001092
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
C21H30O5
56480
N-Hydroxysuccinimide, purum, ≥97.0% (T)
C4H5NO3
S5799
Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt, ≥95% (H-NMR), powder
C16H18N2Na2O14S2