EMAIL THIS PAGE TO A FRIEND
Plant & cell physiology
Functional Characterization of Cucurbitadienol Synthase and Triterpene Glycosyltransferase Involved in Biosynthesis of Mogrosides from Siraitia grosvenorii.
PMID 25759326
Abstract
Mogrosides, the major bioactive components isolated from the fruits of Siraitia grosvenorii, are a family of cucurbitane-type tetracyclic triterpenoid saponins that are used worldwide as high-potency sweeteners and possess a variety of notable pharmacological activities. Mogrosides are synthesized from 2,3-oxidosqualene via a series of reactions catalyzed by cucurbitadienol synthase (CbQ), Cyt P450s (P450s) and UDP glycosyltransferases (UGTs) in vivo. However, the relevant genes have not been characterized to date. In this study, we report successful identification of SgCbQ and UGT74AC1, which were previously predicted via RNA-sequencing (RNA-seq) and digital gene expression (DGE) profile analysis of the fruits of S. grosvenorii. SgCbQ was functionally characterized by expression in the lanosterol synthase-deficient yeast strain GIL77 and was found to accumulate cucurbitadienol as the sole product. UGT74AC1 was heterologously expressed in Escherichia coli as a His-tag protein and it showed specificity for mogrol by transfer of a glucose moiety to the C-3 hydroxyl to form mogroside IE by in vitro enzymatic activity assays. This study reports the identification of CbQ and glycosyltransferase from S. grosvenorii for the first time. The results also suggest that RNA-seq, combined with DGE profile analysis, is a promising approach for discovery of candidate genes involved in biosynthesis of triterpene saponins.