EMAIL THIS PAGE TO A FRIEND
International journal of nanomedicine
In vitro and in vivo effects of graphene oxide and reduced graphene oxide on glioblastoma.
PMID 25759581
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3021
Dicyanocobinamide, ≥93%
C50H72CoN13O8