EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
A new cytotoxic benzophenanthridine isoquinoline alkaloid from the fruits of Macleaya cordata.
PMID 25761206
Abstract
A new cytotoxic benzophenanthridine isoquinoline alkaloid, named cordatine (1), together with one known alkaloid 8-methoxydihydrochelerythrine (2), was isolated from the fruits of Macleaya cordata. The structure of the new compound was elucidated by spectroscopic methods including 1D and 2D NMR, HR-ESI-MS. Both compounds indicated significant cytotoxicity against MCF-7 and SF-268 cell lines.