EMAIL THIS PAGE TO A FRIEND
Journal of Pharmacology and Experimental Therapeutics
VX-509 (decernotinib) is a potent and selective janus kinase 3 inhibitor that attenuates inflammation in animal models of autoimmune disease.
PMID 25762693