EMAIL THIS PAGE TO A FRIEND
Boletin medico del Hospital Infantil de Mexico
[Prokinetic drugs in gastrointestinal motility. Their application in pediatrics].
PMID 2576370
Abstract
In this article, the neurophysiology and neurobiochemistry of the digestive system are reviewed briefly. The pharmacology of the main gastrointestinal prokinetic drugs and their indications, side effects and dosages in pediatric patients are described.