EMAIL THIS PAGE TO A FRIEND
Virology
Novel expression and regulation of TIMP-1 in Epstein Barr virus-infected cells and its impact on cell survival.
PMID 25765004
Abstract
Epstein Barr virus (EBV) uses various strategies to manipulate host cytokine production in favor of the survival of infected B-cells. Microarray and cytokine protein array assays revealed that tissue inhibitor of metalloproteinase-1 (TIMP-1) was significantly up-regulated in EBV-infected primary B cells and maintained in abundance in EBV-immortalized lymphoblastoid cell lines (LCLs). TIMP-1 plays critical roles in extracellular matrix homeostasis and regulates signaling pathways. In this study, we demonstrated that the EBV-encoded immediate early lytic protein, Zta, upregulates mainly TIMP-1 expression by binding to the AP-1 site within the TIMP-1 promoter. Moreover, knockdown of TIMP-1 expression promoted cisplastin and cold shock-induced death of LCLs. This study provides a mechanistic link between EBV-induced TIMP-1 expression and its impact on LCL survival.