EMAIL THIS PAGE TO A FRIEND
Bioorganic & Medicinal Chemistry
Dual inhibition of allosteric mitogen-activated protein kinase (MEK) and phosphatidylinositol 3-kinase (PI3K) oncogenic targets with a bifunctional inhibitor.
PMID 25766633