EMAIL THIS PAGE TO A FRIEND
Indian Journal of Pharmaceutical Sciences
Novel 3,4-Methylenedioxybenzene Scaffold Incorporated 1,3,5-Trisubstituted-2-pyrazolines: Synthesis, Characterization and Evaluation for Chemotherapeutic Activity.
PMID 25767315