EMAIL THIS PAGE TO A FRIEND
Comparative biochemistry and physiology. C, Comparative pharmacology and toxicology
Drug metabolism components in liver microsomes from Hypostomus punctatus, a Brazilian benthic fish (Cascudo).
PMID 2576796
Abstract
1. The major components of hepatic drug biotransformation system were identified in a Brazilian freshwater benthic fish. 2. Cytochrome P-450 difference spectra were obtained adding 0.02 mM phenazine ethosulphate and 2 mM ascorbate to microsomal suspensions. Basal levels of P-450 were high (0.9 nmol/mg of microsomal protein) and were not induced by 3-MC. 3. Microsomal NADPH-cytochrome C reductase activity was determined in presence of 1.3 x 10(-4) M NADPH, 3.3 x 10(-5) M cytochrome C, 1.0 x 10(-4) M EDTA, 66 micrograms of microsomal protein per ml in a 0.3 M Tris-HCl buffer, pH 8.6. Basal levels of NADPH-cytochrome C were 152.7 nmoles/min/mg of microsomal protein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4544
Phenazine ethosulfate, ≥95%
C16H18N2O4S