EMAIL THIS PAGE TO A FRIEND
The Science of the Total Environment
Geochemical markers of soil anthropogenic contaminants in polar scientific stations nearby (Antarctica, King George Island).
PMID 25770449