EMAIL THIS PAGE TO A FRIEND
Talanta
Rapid and selective extraction of multiple macrolide antibiotics in foodstuff samples based on magnetic molecularly imprinted polymers.
PMID 25770599
Abstract
Magnetic molecularly imprinted polymers (MMIPs) were prepared based on surface molecular imprinting using erythromycin (ERY) as template molecule and Fe3O4 nanoparticles as support substrate. The MMIPs possessed high adsorption capacity of 94.1 mg/g for ERY and the imprinting factor was 11.9 indicating good imprinted effect for ERY. Selective evaluation demonstrated favorable selectivity of MMIPs for multiple macrolide antibiotics (MACs). Using MMIPs as adsorptive material, a rapid and convenient magnetic solid-phase extraction (MSPE) procedure was established for simultaneous and selective separation of six MACs in pork, fish and shrimp samples, then the MACs was subjected to high-performance liquid chromatography-ultraviolet (HPLC-UV) analysis. At different fortified concentrations, the extraction recoveries could reach 89.1% and the relative standard deviations were lower than 12.4%. Chromatogram revealed the response signals of MACs in spiked samples were greatly enhanced and matrix interferences were effectively eliminated after treatment with MSPE. The proposed MSPE procedure coupled with HPLC-UV realized selective and sensitive determination of multiple MACs in foodstuff samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
440159
3-(Trimethoxysilyl)propyl methacrylate, 98%
C10H20O5Si
75199
Azithromycin, analytical standard
C38H72N2O12
1046056
Azithromycin, United States Pharmacopeia (USP) Reference Standard
C38H72N2O12 · 2H2O
Y0000306
Azithromycin, European Pharmacopoeia (EP) Reference Standard
C38H72N2O12 · 2H2O
Y0000637
Azithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
C38H72N2O12 · 2H2O
Y0000641
Azithromycin for system suitability, European Pharmacopoeia (EP) Reference Standard
C38H72N2O12 · 2H2O
1046067
Azithromycin Identity, United States Pharmacopeia (USP) Reference Standard
C38H72N2O12
C9742
Clarithromycin, ≥95% (HPLC)
C38H69NO13
PHR1038
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
C38H69NO13
Y0000320
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
C38H69NO13
1134379
Clarithromycin, United States Pharmacopeia (USP) Reference Standard
C38H69NO13
A3487
Clarithromycin, ≥98% (HPLC)
C38H69NO13
Y0000321
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
C38H69NO13
1134390
Clarithromycin Identity, United States Pharmacopeia (USP) Reference Standard
C38H69NO13
14610
Methyl acetoacetate, analytical standard
C5H8O3
R4393
Roxithromycin, ≥90% (HPLC)
C41H76N2O15
R1500000
Roxithromycin, European Pharmacopoeia (EP) Reference Standard
C41H76N2O15