EMAIL THIS PAGE TO A FRIEND
Talanta
Determination of N-nitrosodiethanolamine, NDELA in cosmetic ingredients and products by mixed mode solid phase extraction and UPLC-tandem mass spectrometry with porous graphitic carbon column through systemic sample pre-cleanup procedure.
PMID 25770613
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7632
N-Nitrosodiethanolamine
C4H10N2O3