EMAIL THIS PAGE TO A FRIEND
Toxicology
Participation of divalent cation transporter DMT1 in the uptake of inorganic mercury.
PMID 25772431
Abstract
Mercury (Hg) is found in food in various chemical forms, which differ in terms of accumulation, transport, and toxicity. Although methylmercury (CH3Hg) is the predominant mercury species in the diet, contributed mostly by seafood products, there is also a contribution of inorganic mercury [Hg(II)] from vegetables, cereals, and seafood products. The main pathway for exposure to mercury is oral, and therefore the gastrointestinal mucosa is the first barrier that the contaminant meets when it enters the systemic circulation. However, the transport mechanisms responsible for the process of mercury absorption are not known. The aim of this study is to evaluate the possible participation of divalent metal transporter 1 (DMT1) in Hg(II) intestinal uptake. For this purpose, we have used various complementary approaches. We have studied mercury acquisition in a Saccharomyces cerevisiae strain expressing murine DMT1. Moreover, we have evaluated the effect of a reduction of DMT1 expression in Caco-2 cells, by means of small interfering RNA and of treatment with hepcidin, on mercury uptake and transport. The results show that expression of the transporter DMT1 in yeast produces an increase in Hg(II) accumulation. Furthermore, a decrease in the levels of DMT1 mRNA in Caco-2 cells in various stages of differentiation leads to a reduction in cellular accumulation and apical-basolateral transport of Hg(II). These data point clearly to the mediation of the divalent cation transporter DMT1 in the entry of Hg(II) into the intestinal epithelium.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
266167
Manganese, chips, thickness <2.0 mm, 99%
Mn
266132
Manganese, powder, −325 mesh, ≥99% trace metals basis
Mn
463728
Manganese, powder, ≥99.9% trace metals basis
Mn
GF74177736
Manganese, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF99005193
Manganese, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 98.7%
Mn
GF90559794
Manganese, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF03428625
Manganese, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
Mn
GF31519975
Manganese, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
Mn
GF60677646
Manganese, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
Mn
GF64815239
Manganese, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 98.7%
Mn
GF41418378
Manganese, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
Mn
GF33640860
Manganese, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
Mn
GF85367605
Manganese, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
Mn
GF87981564
Manganese, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 98.7%
Mn
GF53022833
Manganese, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Mn
GF71274258
Manganese, foil, not light tested, 50x50mm, thickness 0.008mm, permanent polyester support, 98.7%
Mn
GF70055524
Manganese, foil, not light tested, 50x50mm, thickness 0.009mm, permanent polyester support, 98.7%
Mn
GF86610539
Manganese, foil, not light tested, 50x50mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn
GF00832081
Manganese, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Mn
GF61911288
Manganese, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF94645233
Manganese, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 98.7%
Mn
GF05710430
Manganese, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF27632055
Manganese, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 98.7%
Mn
GF11021094
Manganese, rod, 10mm, diameter 4.0mm, cast, 99.5%
Mn
GF34573697
Manganese, rod, 35mm, diameter 2.0mm, cast, 99.5%
Mn
GF38495885
Manganese, rod, 50mm, diameter 4.0mm, cast, 99.5%
Mn
GF42039918
Manganese, rod, 50mm, diameter 6.35mm, cast, 99.5%
Mn
GF83932768
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF87355334
Manganese, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF15798730
Manganese, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn