EMAIL THIS PAGE TO A FRIEND
Arch. Pharm. (Weinheim)
New arylpiperazinylalkyl derivatives of 8-alkoxy-purine-2,6-dione and dihydro[1,3]oxazolo[2,3-f]purinedione targeting the serotonin 5-HT1A /5-HT2A /5-HT7 and dopamine D2 receptors.
PMID 25773907