EMAIL THIS PAGE TO A FRIEND
Bratislavske lekarske listy
Topiramate has protective effect on renal injury.
PMID 25773955
Abstract
Topiramate (TPM) decreases tumor necrosis factor-alpha (TNF-α) and oxidative stress. We investigated protective effects of TPM on cell damage in kidney tissue during ischemia-reperfusion (I/R) damage. A total of 30 male Wistar albino rats were divided into three groups: control, I/R, and I/R plus TPM (I/R+TPM). Laparotomy without I/R injury was performed in control group. After laparotomy, cross ligation of infrarenal abdominal aorta was applied for two hours in I/R groups which was followed by two hours of reperfusion. TPM (100 mg/kg/day) was orally administrated to animals in the I/R+TPM group for seven consecutive days before I/R. The I/R group's TNF-α and interleukin-1 beta (IL-1β) levels were significantly higher (1184.2 ± 129.1 pg/mg protein; 413.1 ± 28.8 pg/mg protein, respectively) than those of the control (907.8 ± 113.0 pg/mg protein, p = 0.002; 374.7 ± 23.7 pg/mg protein, p = 0.010, respectively) and I/R+TPM groups (999.5 ± 115.2 pg/mg protein, p < 0.001; 377.9 ± 30.9 pg/mg protein, p = 0.007, respectively). TPM may partially prevent renal damage in rats. The opening of new horizons of this kind of knowledge will help understand the complex challenge in the prevention of renal I/R damage (Tab. 1, Fig. 3, Ref. 42).