EMAIL THIS PAGE TO A FRIEND
PLoS genetics
The Escherichia coli small protein MntS and exporter MntP optimize the intracellular concentration of manganese.
PMID 25774656
Abstract
Escherichia coli does not routinely import manganese, but it will do so when iron is unavailable, so that manganese can substitute for iron as an enzyme cofactor. When intracellular manganese levels are low, the cell induces the MntH manganese importer plus MntS, a small protein of unknown function; when manganese levels are high, the cell induces the MntP manganese exporter and reduces expression of MntH and MntS. The role of MntS has not been clear. Previous work showed that forced MntS synthesis under manganese-rich conditions caused bacteriostasis. Here we find that when manganese is scarce, MntS helps manganese to activate a variety of enzymes. Its overproduction under manganese-rich conditions caused manganese to accumulate to very high levels inside the cell; simultaneously, iron levels dropped precipitously, apparently because manganese-bound Fur blocked the production of iron importers. Under these conditions, heme synthesis stopped, ultimately depleting cytochrome oxidase activity and causing the failure of aerobic metabolism. Protoporphyrin IX accumulated, indicating that the combination of excess manganese and iron deficiency had stalled ferrochelatase. The same chain of events occurred when mutants lacking MntP, the manganese exporter, were exposed to manganese. Genetic analysis suggested the possibility that MntS exerts this effect by inhibiting MntP. We discuss a model wherein during transitions between low- and high-manganese environments E. coli uses MntP to compensate for MntH overactivity, and MntS to compensate for MntP overactivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1127
2-Nitrophenyl β-D-galactopyranoside, ≥98% (enzymatic)
C12H15NO8
73660
2-Nitrophenyl β-D-galactopyranoside, ≥99.0% (HPLC)
C12H15NO8
A3785
5-Aminolevulinic acid hydrochloride, ≥98%
C5H9NO3 · HCl
A7793
5-Aminolevulinic acid hydrochloride, BioReagent, suitable for cell culture, powder, ≥98%
C5H9NO3 · HCl
08339
5-Aminolevulinic acid hydrochloride, ≥97.0% (AT)
C5H9NO3 · HCl
32319
Diethylenetriaminepentaacetic acid, for complexometry, ≥99.0%
C14H23N3O10
463728
Manganese, powder, ≥99.9% trace metals basis
Mn
266132
Manganese, powder, −325 mesh, ≥99% trace metals basis
Mn
266167
Manganese, chips, thickness <2.0 mm, 99%
Mn
GF11021094
Manganese, rod, 10mm, diameter 4.0mm, cast, 99.5%
Mn
GF83932768
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF87355334
Manganese, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF15798730
Manganese, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn
GF20192240
Manganese, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF43172210
Manganese, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 98.7%
Mn
GF41027352
Manganese, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF81243929
Manganese, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
Mn
GF28884549
Manganese, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
Mn
GF40287258
Manganese, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
Mn
GF94007594
Manganese, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
Mn
GF58094635
Manganese, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
Mn
GF05008092
Manganese, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
Mn
GF70110593
Manganese, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%
Mn
GF41613690
Manganese, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 98.7%
Mn
GF53469142
Manganese, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
Mn
GF92898983
Manganese, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%
Mn
GF94542206
Manganese, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn
GF74177736
Manganese, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF24430395
Manganese, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
Mn
GF03428625
Manganese, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
Mn