EMAIL THIS PAGE TO A FRIEND
Journal of medicinal food
Inhibitory Effects of Two Varieties of Tunisian Pomegranate (Punica granatum L.) Extracts on Gastrointestinal Transit in Rat.
PMID 25775227
Abstract
The present study was undertaken to determine whether total and methanol juice extracts of two Tunisian Pomegranate (Punica granatum L.) varieties (Garsi and Gabsi) protect against diarrhea as well as their effects on gastrointestinal transit (GIT) in healthy rats. In this respect, male Wistar rats were used and divided into control- and pomegranate-treated groups. The antidiarrheal activity was evaluated using the castor oil-induced diarrhea method and the GIT was assessed using charcoal meal. Our results showed that total and methanol P. granatum juice extracts produced a significant dose-dependent protection against castor oil-induced diarrhea. Pomegranate extracts and juice also decreased the GIT significantly and dose dependently. Importantly, the Garsi variety appeared to be more effective than the Gabsi variety on these two parameters. These findings suggest that pomegranate extracts have a potent antidiarrheal property in rats confirming their efficiency in the Tunisian traditional medicine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y3125
Yohimbine hydrochloride, ≥98% (HPLC), powder
C21H26N2O3 · HCl