EMAIL THIS PAGE TO A FRIEND
Experimental and Therapeutic Medicine
Synergistic effects of Buyang Huanwu decoction and embryonic neural stem cell transplantation on the recovery of neurological function in a rat model of spinal cord injury.
PMID 25780400