EMAIL THIS PAGE TO A FRIEND
Acta pharmaceutica (Zagreb, Croatia)
Potential antiproliferative effect of isoxazolo- and thiazolo coumarin derivatives on breast cancer mediated bone and lung metastases.
PMID 25781704