EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
Bioactive triterpenoids from Atriplex lasiantha.
PMID 25782461
Abstract
Chromatographic purification of the ethyl acetate soluble fraction from the methanolic extract of Atriplex lasiantha yielded a new triterpenoid, 7β,15α,16β-trihydroxyolean-12-ene-28,30-dioic acid-3-O-β-D-xylopyranoside (1), along with two known triterpenoids, rotundifolioside I (2) and corchorusin B (3). Structures of the compounds 1-3 were elucidated through sophisticated NMR studies and high resolution mass spectrometry. The three isolates (1-3) were evaluated for antibacterial, antioxidant, and antiurease activities. Compound 2 exhibited the best antibacterial activity against Escherichiacoli with IC50 value of 66.25 μg/ml, whereas, all the tested compounds exhibited antioxidant (IC50 values of 68.7-75.4 μg/ml) and antiurease (IC50 values of 25.5-49.3 μg/ml) activities, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
359009
Cerium(IV) sulfate
CeO8S2