EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim An Der Bergstrasse, Germany)
Synthesis and biological evaluation of RGD peptidomimetic-paclitaxel conjugates bearing lysosomally cleavable linkers.
PMID 25784522
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E3132
E-64, protease inhibitor
C15H27N5O5