PloS one
ADAMTS expression in colorectal cancer.
PMID 25786261
Abstract
ADAMTSs are a family of secreted proteinases that share the metalloproteinase domain with matrix metalloproteinases (MMPs). By acting on a large panel of extracellular substrates, they control several cell functions such as fusion, adhesion, proliferation and migration. Through their thrombospondin motifs they also possess anti-angiogenic properties. We investigated whether ADAMTSs participate in colorectal cancer progression and invasion. Their expression was investigated at both mRNA and protein levels. Using RT-PCR, the expression of ADAMTS-1, -4, -5 and ADAMTS-20 was estimated in colorectal tumors of different cancer stage and anatomic site and 3 cell lines of different aggressiveness. An overexpression of ADAMTS-4 and -5 was observed, especially in tissue samples, whereas ADAMTS-1 and -20 were found to be down-regulated. Western blot analysis further supported the RT-PCR findings, revealing in addition the degradation of ADAMTS-1 and -20 in cancer. In situ expression and localization of ADAMTS-1, -4, -5 and -20 was also investigated by immunohistochemical analysis. Our data suggest a positive correlation between ADAMTS-4 and -5 expression and cancer progression, in contrast with the anti-angiogenic members of the family, ADAMTS-1 and -20, which were found to be down-regulated. Our findings support the notion that overexpression of ADAMTS-4 and ADAMTS-5 in colorectal cancer might be a possible invasive mechanism of cancer cells in order to degrade proteoglycans of ECM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01700
Acrylamide, purum, ≥98.0% (GC)
C3H5NO
01696
Acrylamide, for Northern and Southern blotting, powder blend
C3H5NO
08267
Acrylamide, certified reference material, TraceCERT®
C3H5NO
23701
Acrylamide, analytical standard
C3H5NO
01697
Acrylamide solution, 40% in H2O, for molecular biology
C3H5NO
03378
Chloride atomic spectroscopy standard concentrate 10.00 g Cl-, 10.00 g/L, for 1 l standard solution, analytical standard
HCl
54461
HEPES, ≥99.0% (T)
C8H18N2O4S
PHR1428
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
C8H18N2O4S
17933
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization
HCl
17934
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization
HCl
17935
Hydrogen chloride – methanol solution, ~1.25 M HCl, for GC derivatization
HCl
16911
Hydrogen peroxide solution, for ultratrace analysis
H2O2
95321
Hydrogen peroxide solution, ≥30%, for trace analysis
H2O2
95294
Hydrogen peroxide solution, tested according to Ph.Eur.
H2O2
88597
Hydrogen peroxide solution, 3%, for microbiology
H2O2
792519
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
CHNaO3
1613655
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
CHNaO3
71631
Sodium bicarbonate, tested according to Ph.Eur.
CHNaO3