EMAIL THIS PAGE TO A FRIEND
Urologia internationalis
Reduced ING4 Expression Is Associated with the Malignancy of Human Bladder.
PMID 25790869
Abstract
Inhibitor of growth 4 (ING4) is a tumor suppressor. However the role of ING4 in human bladder malignancy is unknown. In this study, ING4 expression in human bladder cancer and its potential effects were studied. ING4 expression in 47 human bladder cancer tissues and paired adjacent normal tissues was detected by Western blotting, quantitative reverse transcription-polymerase chain reaction, and immunohistochemistry. The migration and cell cycle progression of SV-HUC-1 and T24 cells with aberrant ING4 expression were examined. ING4 protein and mRNA were significantly decreased in bladder cancer tissues. ING4 protein level was significantly lower in the group of patients over 50 years of age. ING4 knockdown caused more rapid cell migration and increased the population of SV-HUC-1 and T24 cells in the G2-M phase. Our data suggest a close connection between aberrant ING4 expression and the carcinogenesis of human bladder cells. ING4 may be a potential target for bladder cancer chemotherapy.