EMAIL THIS PAGE TO A FRIEND
Psychopharmacology
Psychopharmacological characterisation of the successive negative contrast effect in rats.
PMID 25791190
Abstract
Successive negative contrast (SNC) describes a change in the behaviour of an animal following a downshift in the quantitative or qualitative value of an expected reward. This behavioural response has been hypothesised to be linked to affective state, with negative states associated with larger and/or prolonged shifts in behaviour. This study has investigated whether different psychopharmacological treatments have dissociable actions on the SNC effect in rats and related these findings to their actions on different neurotransmitter systems and affective state. Animals were trained to perform a nose-poke response to obtain a high-value food reward (four pellets). SNC was quantified during devalue sessions in which the reward was reduced to one pellet. Using a within-subject study design, the effects of acute treatment with anxiolytic, anxiogenic, antidepressant and dopaminergic drugs were investigated during both baseline (four pellets) or devalue sessions (one pellet). The indirect dopamine agonist, amphetamine, attenuated the SNC effect whilst the D1/D2 antagonist, alpha-flupenthixol, potentiated it. The antidepressant citalopram, anxiolytic buspirone and anxiogenic FG7142 had no specific effects on SNC, although FG7142 induced general impairments at higher doses. The α2-adrenoceptor antagonist, yohimbine, increased premature responding but had no specific effect on SNC. Results for the anxiolytic diazepam were mixed with one group showing an attenuation of the SNC effect whilst the other showed no effect. These data suggest that the SNC effect is mediated, at least in part, by dopamine signalling. The SNC effect may also be attenuated by benzodiazepine anxiolytics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7861
Citalopram hydrobromide, ≥98% (HPLC)
C20H21FN2O · HBr
Y3125
Yohimbine hydrochloride, ≥98% (HPLC), powder
C21H26N2O3 · HCl