EMAIL THIS PAGE TO A FRIEND
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation
Low Counts of Plasmacytoid Dendritic Cells after Engraftment Are Associated with High Early Mortality after Allogeneic Stem Cell Transplantation.
PMID 25792371
Abstract
Dendritic cells (DCs) are antigen-presenting cells that drive immune responses and tolerance and are divided in different subsets: myeloid DCs (mDCs: lineage-; HLA-DR+, 11c+), plasmacytoid dendritic cells (pDCs: HLA-DR+, CD123+), and monocyte-derived DCs (moDC: lineage-, 11c+, 16+). After hematopoietic stem cell transplantation (HSCT), low DC counts in the recipients' peripheral blood (PB) have been associated with worse outcomes, but the relevance of DC graft content remains unclear, and there are few data in the setting of unrelated donor HSCT. We evaluated the DC graft content and monitored DC recovery in PB from 111 HSCT recipients (median age, 17 years; range 1 to 74), who received bone marrow (46%), umbilical cord blood (32%), or PB (22%) from unrelated (81%) or related donors (19%). In 86 patients with sustained allogeneic recovery, patients with higher counts of all DC subsets (pDC, mDC, and moDC) 3 weeks after engraftment had lower incidence of nonrelapse mortality (NMR) and acute graft-versus-host disease (aGVHD) and better survival. pDC counts were associated with more striking results: patients with higher pDC counts had much lower incidences of NRM (3% versus 47%, P < .0001), lower incidence of aGVHD (24% versus 67%, P < .0001), and better overall survival (92% versus 45%, P < .0001). In contrast, higher pDC counts in the graft was associated with an increased risk of aGVHD (55% versus 26%, Pxa0= .02). Our results indicate that DC counts are closely correlated with HSCT outcomes and warrant further prospective evaluation and possible early therapeutic interventions to ameliorate severe aGVHD and decrease mortality.