EMAIL THIS PAGE TO A FRIEND
Folia morphologica
Pancreatic remnant fate.
PMID 25792396
Abstract
There is eternal discussion on the best surgical method of pancreatoduodenectomy and reconstruction method. Several different methods of pancreatic stump anastomosis exist. The most popular argument taken into account in the discussion is the frequency of early postoperative complications. Relatively fewer papers analyse the late functional outcome of pancreatic surgery and the method of anastomosis employed. Authors presented short series of 12 patients after pancreatic surgery with analysis of pancreatic remnant morphology and function. Pancreatic remnant volume, pancreatic duct distension and stool elastase-1 test were analysed. There was no correlation of pancreatic exo- or endocrine insufficiency with the volume of pancreatic remnant or the kind of surgery or anastomosis performed.