EMAIL THIS PAGE TO A FRIEND
Life (Basel, Switzerland)
Physical Routes to Primitive Cells: An Experimental Model Based on the Spontaneous Entrapment of Enzymes inside Micrometer-Sized Liposomes.
PMID 25793278