EMAIL THIS PAGE TO A FRIEND
The Journal of hand surgery, European volume
Corrigendum on. Total wrist arthroplasty: a systematic review of the evidence from the last 5 years.
PMID 25794885
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35399
Clencyclohexerol hydrochloride, VETRANAL®, analytical standard
C14H20Cl2N2O2 · HCl