EMAIL THIS PAGE TO A FRIEND
Public health
Tuberculosis report among injection drug users and their partners in Kazakhstan.
PMID 25795015
Abstract
Tuberculosis (TB) is a major threat to global public health. Kazakhstan has the second highest percentage of multidrug-resistant tuberculosis (MDR-TB) cases among incident tuberculosis cases in the world (WHO 2013). A high burden of MDR-TB suggests TB prevention, control, and treatment programs are failing. This study provides an epidemiologic profile of TB among injection drug users (IDUs), a high-risk and chronically underserved population, in Kazakhstan. Cross-sectional study. The authors studied the characteristics and risk environment of IDUs with self-reported previous active TB and their primary sexual partners in Almaty, Kazakhstan. 728 individuals (364 couples) participated in a couple-based study in 2009. 16.75% of participants reported at least one positive TB test (x-ray) in their lifetime. In a multivariable logistic regression adjusting for couple-based sampling, persons with positive TB test were significantly more likely to be older (odds ratio (OR) 7.26, 95% confidence interval (CI): 1.73, 30.43), male (OR 5.53, 95% CI: 2.74, 11.16), have a shorter duration of injection drug use (OR 0.17, 95% CI: 0.04, 0.65), have received high social support from their significant other (OR 2.13, 95% CI: 1.03, 4.40) and more likely (non-significantly) to have been incarcerated (OR 7.03, 95% CI: 0.64, 77.30). Older men with a history of incarceration and recent injection drug use were more likely to have positive TB test in Kazakhstan. Social network support, while potentially positive for many aspects of population health, may increase risk of TB among IDUs in this context. Public health policies that target high-risk populations and their at-risk networks may be necessary to stem the rise of MDR-TB in Central Asia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35399
Clencyclohexerol hydrochloride, VETRANAL®, analytical standard
C14H20Cl2N2O2 · HCl