EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
A molecularly imprinted polymer for the selective solid-phase extraction of dimethomorph from ginseng samples.
PMID 25795323
Abstract
A molecularly imprinted polymer (MIP) was synthesized and evaluated to selectively extract dimethomorph from ginseng samples. Dimethomorph molecularly imprinted polymers with template to monomer molar ratios were contrived and developed via precipitation polymerization employing methacrylic acid as functional monomer, ethylene dimethacrylate as cross-linker and butanone:N-heptane (7:3, v:v)as porogen. The LOD (limit of detection) of this method was 0.002 mg kg(-1), and the LOQ (limit of quantification) was 0.005 mg kg(-1). The different spiked level of ginseng was 0.1 mg kg(-1), 1.0 mg kg(-1), 5.0 mg kg(-1), and the average recovery of dimethomrph was 89.2-91.6%. Under the optimized condition, good linearity was obtained from 0.01 to 5 mg kg(-1) (r(2) ≥ 0.9997) with the relative standard deviations of less than 3.20%. This proposed MISPE-GC procedure eliminated the effect of template leakage on quantitative analysis and could be applied to direct determination of dimethomrph in ginseng samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46027
Dimethomorph, PESTANAL®, analytical standard
C21H22ClNO4