EMAIL THIS PAGE TO A FRIEND
Acta medica Iranica
Association of chronic thromboembolic pulmonary hypertension with hereditary and acquired risk factors for thromboembolism.
PMID 25796019
Abstract
Thrombophilia increases the incidence of pulmonary thrombosis significantly. Various hereditary and acquired factors are known for thromboembolism. The hereditary factors are two common genetic autosomal mutations including factor V leidan mutation and prothrombin gene mutation. A descriptive- analytical design was conducted on 60 patients with thromboembolism who met the inclusion criteria. Two groups with hereditary and acquired risk factors for thromboembolism (group A, B, each 30 people) were evaluated. All the patients of two groups were evaluated in terms of emboli associated with hereditary and acquired risk factors. Association of thromboembolism risk factors' with pulmonary hypertension were studied at the beginning of the study and six months after the treatment then the results of two groups were compared. Among participants, 31 (56.4%) were men.  The mean age of the patients was 44.4±14 years, ranging between 23-75 years. Significant association was observed between the stability of the blood clot in pulmonary vessel, six months after the treatment with genetic risk factors (P=0.03). However, no significant association was between pulmonary hypertension and hereditary and acquired risk factors (P=0.24). Based on the significant association between the hereditary risk factors and pulmonary emboli, by taking special prevention and therapy measurements (e.g. genetic engineering), some pulmonary and mortality complications can be prevented and the patient himself and health care system would benefit from this issue.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35399
Clencyclohexerol hydrochloride, VETRANAL, analytical standard
C14H20Cl2N2O2 · HCl