EMAIL THIS PAGE TO A FRIEND
Developmental biology
Differential levels of Neurod establish zebrafish endocrine pancreas cell fates.
PMID 25797153
Abstract
During development a network of transcription factors functions to differentiate foregut cells into pancreatic endocrine cells. Differentiation of appropriate numbers of each hormone-expressing endocrine cell type is essential for the normal development of the pancreas and ultimately for effective maintenance of blood glucose levels. A fuller understanding of the details of endocrine cell differentiation may contribute to development of cell replacement therapies to treat diabetes. In this study, by using morpholino and gRNA/Cas9 mediated knockdown we establish that differential levels of the basic-helix loop helix (bHLH) transcription factor Neurod are required for the differentiation of distinct endocrine cell types in developing zebrafish. While Neurod plays a role in the differentiation of all endocrine cells, we find that differentiation of glucagon-expressing alpha cells is disrupted by a minor reduction in Neurod levels, whereas differentiation of insulin-expressing beta cells is less sensitive to Neurod depletion. The endocrine cells that arise during embryonic stages to produce the primary islet, and those that arise subsequently during larval stages from the intra-pancreatic duct (IPD) to ultimately contribute to the secondary islets, show similar dependence on differential Neurod levels. Intriguingly, Neurod-deficiency triggers premature formation of endocrine precursors from the IPD during early larval stages. However, the Neurod-deficient endocrine precursors fail to differentiate appropriately, and the larvae are unable to maintain normal glucose levels. In summary, differential levels of Neurod are required to generate endocrine pancreas subtypes from precursors during both embryonic and larval stages, and Neurod function is in turn critical to endocrine function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
309435
3-Methyl-1-butanol, anhydrous, ≥99%
C5H12O
I9392
3-Methylbutanol, BioReagent, for molecular biology, ≥98.5%
C5H12O
59085
3-Methylbutanol, BioUltra, for molecular biology, ≥99.0% (GC)
C5H12O
G3157
Glucagon, from porcine pancreas, powder, γ-irradiated, cell culture tested
C153H225N43O49S
G2044
Glucagon, synthetic, powder, cell culture tested
C153H225N43O49S
G1774
Glucagon, ≥95% (HPLC), powder, synthetic
C153H225N43O49S
W205702
Isoamyl alcohol, ≥98%, FG
C5H12O
W205710
Isoamyl alcohol, natural, ≥98%, FG
C5H12O
P9346
Liquified Phenol, meets USP testing specifications, ≥89.0%
C6H6O
G2654
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid
P1037
Phenol, for molecular biology
C6H6O
P5566
Phenol, BioXtra, ≥99.5% (GC)
C6H6O
328111
Phenol, unstabilized, purified by redistillation, ≥99%
C6H6O
242322
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
C6H6O
77607
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
C6H6O
33517
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
C6H6O
16016
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
C6H6O
16017
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
C6H6O
W322318
Phenol, ≥99%
C6H6O
W322340
Phenol, natural, 97%, FG
C6H6O
P4557
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
C6H6O
P4682
Phenol solution, BioReagent, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
C6H6O
S9129
Somatostatin, ≥97% (HPLC), powder
C76H104N18O19S2
S1763
Somatostatin, powder, BioReagent, suitable for cell culture
C76H104N18O19S2
S0885
Somatostatin, powder, γ-irradiated, BioXtra, suitable for cell culture
C76H104N18O19S2