PLoS ONE
Role of the sympathetic nervous system in carbon tetrachloride-induced hepatotoxicity and systemic inflammation.
PMID 25799095
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
02671
Carbon tetrachloride, analytical standard
CCl4