EMAIL THIS PAGE TO A FRIEND
Journal of Mass Spectrometry : Jms
Studies on the collision-induced dissociation of adipoR agonists after electrospray ionization and their implementation in sports drug testing.
PMID 25800023