EMAIL THIS PAGE TO A FRIEND
Nature Communications
Xanthine oxidoreductase regulates macrophage IL1? secretion upon NLRP3 inflammasome activation.
PMID 25800347
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A10809
4′-Hydroxy-3′-methoxyacetophenone, 98%
C9H10O3
W508454
Acetovanillone, ≥98%, FG
C9H10O3
A8003
Allopurinol, xanthine oxidase inhibitor
C5H4N4O
A0350000
Allopurinol, European Pharmacopoeia (EP) Reference Standard
C5H4N4O
PHR1377
Allopurinol, Pharmaceutical Secondary Standard; Certified Reference Material
C5H4N4O
1013002
Allopurinol, United States Pharmacopeia (USP) Reference Standard
C5H4N4O
60060
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, ≥99.0% (T)
AlKO8S2 · 12H2O
31242
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
AlKO8S2 · 12H2O
12623
Aluminum potassium sulfate dodecahydrate, puriss., meets analytical specification of Ph. Eur., BP, 99.0-100.5% (calc. on dry substance)
AlKO8S2 · 12H2O
237086
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
AlKO8S2 · 12H2O
G9005
D-Gluconic acid sodium salt, ≥99%
C6H11NaO7
64188
D-Gluconic acid sodium salt, certified reference material, TraceCERT®
C6H11NaO7
O111
Octoclothepin maleate salt, solid
C19H21ClN2S · C4H4O4
G4500
Potassium D-gluconate, ≥99%
C6H11KO7
P1847
Potassium gluconate, meets USP testing specifications, anhydrous
C6H11KO7
PHR1130
Potassium gluconate, Pharmaceutical Secondary Standard; Certified Reference Material
C6H11KO7
1550001
Potassium gluconate, United States Pharmacopeia (USP) Reference Standard
C6H11KO7
S2054
Sodium gluconate, meets USP testing specifications
C6H11NaO7