EMAIL THIS PAGE TO A FRIEND
Cancer genetics
Emergence of clonal hematopoiesis in the majority of patients with acquired aplastic anemia.
PMID 25800665
Abstract
Acquired aplastic anemia (aAA) is a nonmalignant disease caused by autoimmune destruction of early hematopoietic cells. Clonal hematopoiesis is a late complication, seen in 20-25% of older patients. We hypothesized that clonal hematopoiesis in aAA is a more general phenomenon, which can arise early in disease, even in younger patients. To evaluate clonal hematopoiesis in aAA, we used comparative whole exome sequencing of paired bone marrow and skin samples in 22 patients. We found somatic mutations in 16 patients (72.7%) with a median disease duration of 1xa0year; of these, 12 (66.7%) were patients with pediatric-onset aAA. Fifty-eight mutations in 51 unique genes were found primarily in pathways of immunity and transcriptional regulation. Most frequently mutated was PIGA, with seven mutations. Only two mutations were in genes recurrently mutated in myelodysplastic syndrome. Two patients had oligoclonal loss of the HLA alleles, linking immune escape to clone emergence. Two patients had activating mutations in key signaling pathways (STAT5B (p.N642H) and CAMK2G (p.T306M)). Our results suggest that clonal hematopoiesis in aAA is common, with two mechanisms emerging-immune escape and increased proliferation. Our findings expand conceptual understanding of this nonneoplastic blood disorder. Future prospective studies of clonal hematopoiesis in aAA will be critical for understanding outcomes and for designing personalized treatment strategies.